Acrodisc One™ Syringe Filters With WWPTFE Membrane
Acrodisc One™ syringe filters with wwPTFE membrane are the premium universal filter for both organic and aqueous solutions in HPLC and UHPLC sample prep.
View Products
Request Sample
  • Versatile wwPTFE (water wettable polytetrafluoroethylene) membrane for aqueous and aggressive organic solvent-based solutions
  • Eliminates the membrane selection process by replacing it with a universal membrane
  • Reduces time for method validation with higher analyte recoveries and lower extractables
  • Extends HPLC column life up to 52 times
  • Certified low levels of UV-absorbing extractables for accurate analysis for HPLC/UHPLC
  • Easy filtration of particulate-laden samples with available GxF multi-layer pre-filter
  • 13 mm Acrodisc syringe filter, with minispike configuration, offers low hold-up and easy filtration into autosampler vials
Applications
  • Ideal for filtering HPLC/UHPLC samples and mobile phases
  • Provides two to four times the throughput of standard pre-filter devices for extremely viscous samples