Cleanroom dispensing & disposal unit
This stand alone dispensing and disposal unit is a great combination in any existing or new cleanroom gowning area.
  • Modern, aesthetic and attractive design
  • Suitable for all clean industries
  • Modular, easy to install
  • Construction material: Stainless steel 304L with electropolished finish
  • Does not need to be connected to a wall
Description WxDxH (mm) Cat. No. Promo price Quantity
Dispenser with 4 compartments 800x205x600 134-0504E
Disposal cabinet 800x430x900 134-0486E
Frame connecting dispenser to cabinet  - 134-0487E