Evaporation
Rotary Evaporator
Used for the efficient and gentle removal of solvents from samples by evaporation, under reduced atmospheric pressure, thus allowing the solvents to evaporate faster at low temperatures.
Physical or thermal separation is a common technique in many chemistry laboratories. VWR have an extensive range of equipment for automating these techniques including the great value for money joint IKA/VWR Collection rotary evaporator plus all the glassware you need to create your own combinations.   
Product selection
Evaporators
Variety of models to suit your application perfectly...
Pumps
A range of pumps for all uses...
Chillers
A range of chillers optimised for use with rotary evaporators ensuring constant, reproducible cooling conditions...