Moisture Analysis
VWR offer a choice of infrared heating, halogen, metal, ceramic or coiled quartz element moisture analysers for routine and precision measurements.
  • Infrared heating - no glass (MB23), halogen for fast results (MB25), heating elements with glass or aluminium panels (MA35)
  • High precision moisture analysers: Halogen (MB35, MB45, MA 100), ceramic or coiled quartz elements with (MA100 and MA150)
  • A range of verified versions (OCE) for use in legal metrology is available
Product selection
Moisture Balances
A choice of moisture balances...
Weighing & Drying Dishes
Sturdy aluminium dishes compatible with most moisture balances and dryers. Oil-free and tapered for easy stacking...