Would you like to log in again?
Automated microbial detection system, TECTA™
Supplier: PATHOGEN DETECTION SYSTEMS INC
The TECTA™ B16 provides on-site water testing solution designed to detect E. coli and total coliforms. It is suitable for use with drinking water, waste water, reclaim water, bottled water and ice packing plants. The system consists of the TECTA™ automated microbiology system and the TECTAlert™ test cartridge. The networking capabilities allow for immediate notification and early warning of positive sample results as soon as they occur. The front touch-screen display interface (17,7 cm) controls all functions and an optional USB port for the keyboard. This self-contained bench top device contains 16 incubation chambers. The patented polymer Partition™ test technology senses fluorescent indicator of enzyme activity of the target organism, excludes water sample from optical path for high turbidity resistance. Manual sample acquisition with Direct-to-Test™ capability eliminates sample handling and mixing of reagents.
  • Fully automated, accurate, sensitive and reliable results
  • Continuous sample monitoring coupled with automated sample interpretation
  • Two modes of function: Run mode (incubation and interpretation in 2 - 18 hours) and Read mode (Interpretation of pre-incubated samples in 60 seconds)
  • Results can be forwarded to any device including computers, tablets and smart phones
Detection time in 100 ml sample: <1 CFU (within 18 h); 102 CFU (within 12 h); 104 CFU (within 8 h); 106 CFU (within 2 - 4 h).
Ordering Information: The system includes 16 simultaneous tests (230 V). TECTAlert™ CCA, combined E. coli and total coliform test 100 ml, sample Direct-to-Test™ pre-filled with media and reagents (48 tests/box).
UL Listed, CE compliant and RoHS compliant.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Pk VWR Catalog Number Quantity
Automated microbial detection system, TECTA™, 230 V, 480×623×344 mm 1 PDSYTECTAB16A230
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty