Rockers, Vari-Mix™ and Speci-Mix™
Supplier: Thermo Scientific
Vari-Mix™
BARNM48720-33QEA 1470 SGD
BARNM71010-33Q BARNM48720-33Q
Rockers, Vari-Mix™ and Speci-Mix™
Shakers Rotating/Rocking Shakers
Vari-Mix™ and Speci-Mix™ test tube rockers have smooth rocking action helps to obtain the uniform specimen suspension. These compact and precision-controlled platform mixers can hold tubes of Ø 10 to 30 mm and are ideal for blood collection tubes.
  • Choose from fixed or adjustable speeds
  • Autoclavable white silicone rubber pad enhances viewing
Reversible pad provides one side for 110 mm long capped tubes and other side for longer tubes. These rockers can be used in a noncondensing incubator or cold room with ambient operating conditions of 17 to 27 °C and 20 to 80% RH non condensing.
Delivery information: Includes white silicon rubber friction grip pad for holding tubes in place.