Would you like to log in again?
Cell culture flasks, treated, Nunc™
Supplier: Thermo Scientific
Nunc®
PS, sterile
  • Quick volume estimation and sample identification; includes bold printed graduationsfrom 100 to 500 mL, in 50 mL graduations and large writing patch
  • Maintains optimal sterility during growth and passage; filter cap incorporates a 0.2 µm pore size hydrophobic membrane that allows gas exchange while maintaining sterility
  • Easy to stack; prominent stacking feet on upper surface facilitate reliable stacking in incubators and under your culture hood
This total volume flasks provides a large culture area of single-layer flasks and is cell culture treated to promote cell attachment and expanded growth. Durable construction, friendly design; blow-molded, one-piece design with straight neck and molded grip notches that make this 1,900mL flask easy to hold. Non-pyrogenic.
ORDER
Expand All / Collapse All
Capacity Cap Growth area Neck Pk VWR Catalog Number Quantity
1900 ml Filter 300 cm² straight 12 NUNC132098
Supplier Article Number:: 132098
1900 ml Vent/close 300 cm² straight 12 NUNC132097
Supplier Article Number:: 132097
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty