Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium hydrate
, [Ba(TMHD)2]
ALFA14539.06EA
0
SGD
ALFA14539.06
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate ≥98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate
MOCVD of superconductors
Formula:
C₂₂H₃₈BaO₄·H₂O MW: 521,877 g/mol Boiling Pt: 225 °C (0,3 mmHg) Melting Pt: 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200255 CAS Number: 17594-47-7 UN: 1564 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...