Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) hydrate ≥98%

Supplier: Alfa Aesar
Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium hydrateMg(TMHD)2Magnesium-DPM hydrate

ALFA38599.03EA 136 SGD InStock
ALFA38599.03
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) hydrate ≥98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')magnesium hydrate


Formula: C₂₂H₃₈MgO₄·xH₂O
MW: 390,85 g/mol (anhydrous)
Melting Pt: 135…150 °C …H₂O
MDL Number: MFCD00167208
CAS Number: 339527-87-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty