Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lead(II)

Supplier: Alfa Aesar
Danger

Synonyms: Pb(TMHD)2Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)

ALFA39397.06EA 207 SGD
ALFA39397.06 ALFA39397.03
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lead(II)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lead(II)
Formula: C₂₂H₃₈O₄Pb
Boiling Pt: 134 °C (0,1 mmHg) (subl.)
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00070468
CAS Number: 21319-43-7
UN: 3467
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR