Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III)

Supplier: Alfa Aesar
Warning

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)erbium(III)Er(TMHD)3

ALFA19602.06EA 357 SGD InStock
ALFA19602.06 ALFA19602.03
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III)
Formula: C₃₃H₅₇ErO₆
MW: 717,061 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00015705
CAS Number: 35733-23-4
EINECS: 000-000-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR