Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III) ≥95%

Supplier: Alfa Aesar
Warning

Synonyms: Sc(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III)

ALFA42787.02EA 146 SGD
ALFA42787.02 ALFA42787.04
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III) ≥95%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III)

Intermediate for scandium oxide ceramics

Formula: C₃₃H₅₇O₆Sc
Boiling Pt: 275 °C (dec.)
Melting Pt: 150…152 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00169875
CAS Number: 15492-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR