Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) ≥98%

Supplier: Alfa Aesar
Warning
Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)Y(TMHD)3

ALFA14480.06EA 569 SGD InStock
ALFA14480.06 ALFA14480.03
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) ≥98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III)


Formula: C₃₃H₅₇O₆Y
MW: 638,708 g/mol
Boiling Pt: 95 °C (0,05 mmHg)
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00015713
CAS Number: 15632-39-0
EINECS: 000-000-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty