Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) ≥99.9% (metals basis), recrystallized
Supplier: Thermo Scientific
Synonyms:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)
, Y(TMHD)3
ALFA44003.03EA
0
SGD
ALFA44003.03
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) ≥99.9% (metals basis), recrystallized
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III)
Formula:
C₃₃H₅₇O₆Y MW: 638,708 g/mol Melting Pt: 170…173 °C |
MDL Number:
MFCD00015713 CAS Number: 15632-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...