Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')niobium(IV)

Supplier: Alfa Aesar

Synonyms: Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV)Nb(TMHD)4

ALFA39459.06EA 604 SGD
ALFA39459.06 ALFA39459.03
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')niobium(IV)
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')niobium(IV)
Formula: C₄₄H₇₆NbO₈
Boiling Pt: 325 °C (dec.)
Melting Pt: 219…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00145379
CAS Number: 41706-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR