Hafnium(IV) acetylacetonate ≥97%
|
Warning
|
Synonyms:
Hafnium(IV) 2,4-pentanedionate
ALFA42463.06EA
298
SGD
ALFA42463.06
ALFA42463.03
ALFA42463.14
Hafnium(IV) acetylacetonate ≥97%
Hafnium (IV) acetylacetonate
Formula:
C₂₀H₂₈O₈Hf Melting Pt: 193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044997 CAS Number: 17475-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...