Reinecke's salt

Supplier: Panreac Química SLU

PANR134220.1606EA 271 SGD
PANR134220.1606 PANR134220.1605
Reinecke's salt
Reinecke's salt
Formula: C₄H₁₀CrN₇S₄
MDL Number: MFCD00151654
CAS Number: 13573-16-5
EINECS: 237-003-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR