3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3,5-Dimethylpiperidine
ALFAB23435.22EA
344
SGD
ALFAB23435.22
ALFAB23435.14
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) ≥97%
3,5-Dimethylpiperidin (mixture of cis- and trans-isomers)
Formula:
C₇H₁₅N MW: 113,20 g/mol Boiling Pt: 142…146 °C Density: 0,830 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005996 CAS Number: 35794-11-7 EINECS: 252-730-6 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...