(±)-1-Benzyl-3-hydroxypiperidine ≥99%

Supplier: Alfa Aesar
Warning

Synonyms: 1-Benzyl-3-piperidinol1-Benzyl-3-hydroxypiperidine

ALFAL16375.06EA 503 SGD
ALFAL16375.06 ALFAL16375.03
(±)-1-Benzyl-3-hydroxypiperidine ≥99%
(±)-1-Benzyl-3-hydroxypiperidine
Formula: C₁₂H₁₇NO
MW: 191,27
Boiling Pt: 140…142 °C (6 mmHg)
Density: 1,056
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00023734
CAS Number: 14813-01-5
EINECS: 238-881-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR