Fmoc-L-phenylalanine

Supplier: Panreac Química SLU

Synonyms: N-α-Fmoc-L-PhenylalanineN-(9-Fluorenylmethoxycarbonyl)-L-phenylalanineN-Fmoc-L-phenylalanineFmoc-Phe-OH

PANR15B112.1604EA 101 SGD
PANR15B112.1604 PANR15B112.1606
Fmoc-L-phenylalanine
Fmoc-L-phenylalanine
Formula: C₂₄H₂₁NO₄
MW: 387,4 g/mol
MDL Number: MFCD00037128
CAS Number: 35661-40-6
EINECS: 252-661-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR