Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5,6-Dimethyl-2,1,3-benzothiadiazole ≥97%

Supplier: Alfa Aesar
Warning

ALFAB20755.06EA 820 SGD InStock
ALFAB20755.06 ALFAB20755.03
5,6-Dimethyl-2,1,3-benzothiadiazole ≥97%
5,6-Dimethyl-2,1,3-benzothiadiazole


Formula: C₈H₈N₂S
MW: 164,23
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00174277
CAS Number: 1887-60-1
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty