Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methylbenzothiazolium iodide ≥98%

Supplier: Alfa Aesar
Warning

ALFAL04837.30EA 549 SGD InStock
ALFAL04837.30
3-Methylbenzothiazolium iodide ≥98%
3-Methylbenzothiazolium iodide


Formula: C₈H₈INS
MW: 277,13
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00050297
CAS Number: 2786-31-4
EINECS: 220-505-1
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty