2-Methyl-4-phenylthiazol ≥97%

Supplier: Alfa Aesar

ALFAH51728.06EA 430 SGD InStock
ALFAH51728.06 ALFAH51728.03
2-Methyl-4-phenylthiazol ≥97%
2-Methyl-4-phenylthiazol
Formula: C₁₀H₉NS
MW: 175,25
Melting Pt: 51…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00489998
CAS Number: 1826-16-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR