4-Methyl-2-phenylthiazol ≥98%

Supplier: Alfa Aesar

ALFAA19572.03EA 49.6 SGD
ALFAA19572.03 ALFAA19572.06
4-Methyl-2-phenylthiazol ≥98%
4-Methyl-2-phenylthiazol
Formula: C₁₀H₉NS
Storage Temperature: Ambient
MDL Number: MFCD00272328
CAS Number: 1826-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR