4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid ≥97%

Supplier: Alfa Aesar
Warning

ALFAH51711.06EA 477 SGD InStock
ALFAH51711.06 ALFAH51711.03
4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid ≥97%
4-Methyl-2-(2-pyridinyl)-5-thiazolecarboxylic acid
Formula: C₁₀H₈N₂O₂S
MW: 220,25
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD01316364
CAS Number: 34418-48-9
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR