2-(2-Pyridinyl)-4-thiazolecarboxylic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
ALFAH51851.06EA
0
SGD
ALFAH51851.06
2-(2-Pyridinyl)-4-thiazolecarboxylic acid ≥97%
2-(2-Pyridinyl)-4-thiazolecarboxylic acid
Formula:
C₉H₆N₂O₂S MW: 206,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375352 CAS Number: 115311-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...