2-Amino-4-(4-pyridyl)thiazole 97%

Supplier: Alfa Aesar
Warning

ALFAH52238.03EA 264 SGD
ALFAH52238.03 ALFAH52238.06
2-Amino-4-(4-pyridyl)thiazole 97%
2-Amino-4-(4-pyridyl)thiazole
Formula: C₈H₇N₃S
Storage Temperature: Ambient
MDL Number: MFCD00490009
CAS Number: 30235-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR