3-Bromo-6-chlorochromone 96%

Supplier: Alfa Aesar

ALFAH32041.04EA 343 SGD
ALFAH32041.04 ALFAH32041.MD
3-Bromo-6-chlorochromone 96%
3-Bromo-6-chlorochromone
Formula: C₉H₄BrClO₂
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD02093123
CAS Number: 73220-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR