Tricyclohexylphosphine oxide

Supplier: Thermo Scientific
Danger

ALFA30387.03EA 97.3 SGD
ALFA30387.03 ALFA30387.14 ALFA30387.06
Tricyclohexylphosphine oxide
Tricyclohexylphosphine oxide
Formula: C₁₈H₃₃OP
MW: 296,42 g/mol
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00014298
CAS Number: 13689-19-5
UN: 3464
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR