2-Amino-4-(3-bromophenyl)pyrimidine 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(3-Bromophenyl)pyrimidin-2-amine
ALFAH34024.03EA
0
SGD
ALFAH34024.03
2-Amino-4-(3-bromophenyl)pyrimidine 95%
2-Amino-4-(3-bromophenyl)pyrimidine
Formula:
C₁₀H₈BrN₃ MW: 250,10 g/mol Melting Pt: 203…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD01940446 CAS Number: 392307-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...