2-Amino-4-(4-bromophenyl)pyrimidine 98%

Supplier: Alfa Aesar
Warning

ALFAH33561.06EA 525 SGD
ALFAH33561.06 ALFAH33561.03
2-Amino-4-(4-bromophenyl)pyrimidine 98%
2-Amino-4-(4-bromophenyl)pyrimidine
Formula: C₁₀H₈BrN₃
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD02317272
CAS Number: 392326-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR