2-Acetamido-5-chlorothiazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-(5-Chlorothiazol-2-yl)acetamide
ALFAL17151.03EA
0
SGD
ALFAL17151.03
ALFAL17151.06
2-Acetamido-5-chlorothiazole ≥97%
2-Acetamido-5-chlorothiazole
Formula:
C₅H₅ClN₂OS MW: 176,63 g/mol Melting Pt: 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318098 CAS Number: 20256-39-7 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...