1,2:3,4-Diepoxybutane 95%

Supplier: Alfa Aesar
Danger

Synonyms: Butadiene diepoxide1,3-Butadiene diepoxide

ALFAL03919.06EA 193 SGD
ALFAL03919.06
1,2:3,4-Diepoxybutane 95%
1,2:3,4-Diepoxybutane
Formula: C₄H₆O₂
MW: 86,09
Boiling Pt: 56…58 °C (25 mmHg)
Melting Pt: –19 °C
Density: 1,112
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005120
CAS Number: 1464-53-5
EINECS: 215-979-1
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR