D-(-)-Penicillamine

Supplier: EDQM

Synonyms: H-D-Pen-OH

EDQMP0310000EA 368 SGD InStock
EDQMP0310000
D-(-)-Penicillamine
D(-)-Penicillamine
Formula: C₅H₁₁NO₂S
MW: 149,2 g/mol
MDL Number: MFCD00064302
CAS Number: 52-67-5
EINECS: 200-148-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR