2-Trimethylsilyl-1-ethylboronic acid pinacol ester ≥97%
Supplier: Thermo Scientific
ALFAL19534.MDEA
0
SGD
ALFAL19534.MD
ALFAL19534.03
2-Trimethylsilyl-1-ethylboronic acid pinacol ester ≥97%
2-Trimethylsilyl-1-ethylboronic acid pinacol ester
Formula:
C₁₁H₂₅BO₂Si MW: 228,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788725 CAS Number: 165904-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...