Trimethylsilyl bromoacetate ≥98%

Supplier: Alfa Aesar
Danger

ALFAL00961.14EA 129 SGD
ALFAL00961.14
Trimethylsilyl bromoacetate ≥98%
Trimethylsilyl bromoacetate
Formula: C₅H₁₁BrO₂Si
MW: 211,13
Boiling Pt: 68…69 °C (17 mmHg)
Density: 1,291
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000187
CAS Number: 18291-80-0
EINECS: 242-165-3
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR