2'-(Methylsulphonyl)acetanilide 96%

Supplier: Alfa Aesar
Warning

ALFAH26907.14EA 810 SGD
ALFAH26907.14 ALFAH26907.03 ALFAH26907.06
2'-(Methylsulphonyl)acetanilide 96%
2'-(Methylsulphonyl)acetanilide
Formula: C₉H₁₁NO₃S
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD01464107
CAS Number: 20628-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR