4'-(Methylsulphonyl)acetanilide 96%

Supplier: Alfa Aesar
Warning

ALFAH26922.14EA 345 SGD
ALFAH26922.14 ALFAH26922.06
4'-(Methylsulphonyl)acetanilide 96%
4'-(Methylsulphonyl)acetanilide
Formula: C₉H₁₁NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00025068
CAS Number: 22821-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR