Ethyl phenoxyacetate

Supplier: Panreac Química SLU

PANR15C003.1608EA 465 SGD
PANR15C003.1608 PANR15C003.1606
Ethyl phenoxyacetate
Ethyl phenoxyacetate
Formula: C₁₀H₁₂O₃
MW: 180,20
MDL Number: MFCD00026895
CAS Number: 2555-49-9
EINECS: 219-867-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR