2,2,2-Trifluoro-4'-methoxyacetanilide ≥97%

Supplier: Alfa Aesar
Danger

ALFAH32011.03EA 112 SGD
ALFAH32011.03 ALFAH32011.06
2,2,2-Trifluoro-4'-methoxyacetanilide ≥97%
2,2,2-Trifluoro-4'-methoxyacetanilide
Formula: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00452588
CAS Number: 332-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR