3-Amino-3-(2-chlorophenyl)propionic acid 97%

Supplier: Alfa Aesar

ALFAL19796.03EA 199 SGD
ALFAL19796.03 ALFAL19796.MD
3-Amino-3-(2-chlorophenyl)propionic acid 97%
3-Amino-3-(2-chlorophenyl)propionic acid
Formula: C₉H₁₀ClNO₂
Melting Pt: ∼ 223 °C (dec.)
MDL Number: MFCD01871299
CAS Number: 68208-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR