Potassium-2-formylphenyltrifluoroborate ≥97%

Supplier: Alfa Aesar
Warning

ALFAL17968.03EA 203 SGD
ALFAL17968.03
Potassium-2-formylphenyltrifluoroborate ≥97%
Potassium 2-formylphenyltrifluoroborate
Formula: C₇H₅BF₃KO
Storage Temperature: Ambient
MDL Number: MFCD02093979
CAS Number: 192863-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR