Potassium-3-formylphenyltrifluoroborate ≥97%

Supplier: Alfa Aesar
Warning

ALFAH28125.06EA 256 SGD
ALFAH28125.06 ALFAH28125.03
Potassium-3-formylphenyltrifluoroborate ≥97%
Potassium 3-formylphenyltrifluoroborate
Formula: C₇H₅BF₃KO
MW: 212,02
Melting Pt: ∼ 200 °C
Storage Temperature: Ambient
MDL Number: MFCD07784378
CAS Number: 871231-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR