Potassium-4-(aminocarbonyl)phenyltrifluoroborate ≥96%

Supplier: Alfa Aesar
Warning

Synonyms: Potassium 4-carbamoylphenyltrifluoroborate

ALFAH53054.06EA 367 SGD InStock
ALFAH53054.06 ALFAH53054.03
Potassium-4-(aminocarbonyl)phenyltrifluoroborate ≥96%
Potassium 4-(aminocarbonyl)phenyltrifluoroborate
Formula: C₇H₆BF₃KNO
MW: 227,03
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115758
CAS Number: 850623-63-1
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR