Potassium-4-(phenylaminocarbonyl)phenyltrifluoroborate 97%

Supplier: Alfa Aesar
Warning

ALFAH51150.03EA 59.4 SGD
ALFAH51150.03 ALFAH51150.06
Potassium-4-(phenylaminocarbonyl)phenyltrifluoroborate 97%
Potassium 4-(phenylaminocarbonyl)phenyltrifluoroborate
Formula: C₁₃H₁₀BF₃KNO
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD11977719
CAS Number: 912350-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR