Potassium-3-carboxyphenyltrifluoroborate 95%

Supplier: Alfa Aesar
Warning

ALFAH51885.06EA 292 SGD
ALFAH51885.06 ALFAH51885.03
Potassium-3-carboxyphenyltrifluoroborate 95%
Potassium 3-carboxyphenyltrifluoroborate
Formula: C₇H₅BF₃KO₂
Storage Temperature: Ambient
MDL Number: MFCD07784423
CAS Number: 850313-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR