Potassium-4-carboxyphenyltrifluoroborate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Potassium (4-carboxyphenyl)trifluoroborate
ALFAH51888.03EA
84.7
SGD
ALFAH51888.03
Potassium-4-carboxyphenyltrifluoroborate 95%
Potassium 4-carboxyphenyltrifluoroborate
Formula:
C₇H₅BF₃KO₂ MW: 228,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115737 CAS Number: 850623-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...