Potassium-4-(ethylthio)phenyltrifluoroborate 96%

Supplier: Alfa Aesar
Warning

ALFAH52677.03EA 115 SGD
ALFAH52677.03
Potassium-4-(ethylthio)phenyltrifluoroborate 96%
Potassium 4-(ethylthio)phenyltrifluoroborate
Formula: C₈H₉BF₃KS
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115771
CAS Number: 850623-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR