3-Bromo-4-isopropoxy-5-methoxybenzonitrile ≥98%
Supplier: Thermo Scientific
|
Danger
|
ALFAH50615.03EA
457
SGD
ALFAH50615.03
3-Bromo-4-isopropoxy-5-methoxybenzonitrile ≥98%
3-Bromo-4-isopropoxy-5-methoxybenzonitrile
Formula:
C₁₁H₁₂BrNO₂ MW: 270,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02256549 CAS Number: 515848-62-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...