Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dibromo-5-(trifluoromethoxy)anisole ≥97%

Supplier: Alfa Aesar
Warning

ALFAH33545.03EA 103 SGD InStock
ALFAH33545.03 ALFAH33545.06
2,4-Dibromo-5-(trifluoromethoxy)anisole ≥97%
2,4-Dibromo-5-(trifluoromethoxy)anisole


Formula: C₈H₅Br₂F₃O₂
MW: 349,93
Storage Temperature: Ambient
CAS Number: 1373920-68-3
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty